missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S. Origin)

Product Code. 12604217
Change view
Click to view available options
Processing:
Heat Inactivated
None
Unit Size:
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Processing unitSize
12604217 Heat Inactivated 50mL
11596531 None 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12604217 Supplier Cytiva Supplier No. SH30071.01HI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enhance growth of cells. Tested for endotoxin, hemoglobin, and additional biochemical assays.

  • Provides important proteins, hormones, growth factors, metabolites, and nutrients for your cell culture process
  •  Produced using true pool technology, which minimizes bottle-to-bottle variability
  • Assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC), percent iron saturation, glucose, and IgG
  • U.S. sourced
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Saturation,Glucose,IgG,Gamma Globulin
    Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
    Country of Origin United States
    Product Type Fetal Bovine Serum
    Quantity 50 mL
    Sterility Sterile
    Sterilization Method Sequential Triple-filtered, 100 nm
    Grade Characterized
    Processing Heat Inactivated
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 25 EU/mL
    Heat Inactivated Yes
    Hemoglobin Concentration ≤ 25 mg/dL
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.