missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, New Zealand Origin
Click to view available options
Quantity:
50 mL
500 mL
Processing:
Embryonic Stem Cell Screened
HI, Irr - 25 to 40 doses
Irradiated
Irradiated (30 to 40 kGy)
Mesenchymal Stem Cell Screened
Unit Size:
500mL
50mL
Description
- Meets the requirements of most users
- Filtered through triple 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Specifications
| Certifications/Compliance | Certificate of Suitability No. R1-CEP 2001-211 |
| Country of Origin | New Zealand |
| Form | Serum |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Processing | Mesenchymal Stem Cell Screened |
| Volume (Metric) | 500 mL |
| Endotoxin Concentration | ≤ 20 EU/mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction