missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)

Enhance growth of cells with New Zealand sourced Characterized FBS.

Brand:  Cytiva SH30406.02IR30-45

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 16291112

  • £985.00 / 500mL
Estimated Shipment: 20-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Carefully collected, processed, and filtered in our own New Zealand facility
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • Greater security against bovine disease, since New Zealand has the fewest reported bovine diseases in the world
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles using true pool technology
  • Specifications

    Specifications

    Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen
    500 mL
    Sequential-filtered, 100 nm
    Irradiated, 30 to 45 doses
    ≤ 20 EU/mL
    ≤ 25 mg/dL
    USDA Inspected
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Sterile
    Characterized
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    No
    Yes
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers