missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)
Enhance growth of cells with New Zealand sourced Characterized FBS.
Brand: Cytiva SH30406.02IR
Code : 85
Additional Details : Weight : 0.50000kg
Description
Specifications
Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen | |
Fetal Bovine Serum | |
Sterile | |
None | |
≤ 20 EU/mL | |
≤ 25 mg/dL | |
USDA Inspected |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
500 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Yes |