missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Characterized Fetal Bovine Serum, Australia Origin
Enhances the growth of cells
Brand: Cytiva (Formerly GE Healthcare Life Sciences) SH30084.02IR
Code : 85
Additional Details : Weight : 0.10000kg
Description
- Considered to be free of BSE and FMD
- USDA safety-tested
- Tested for endotoxin and hemoglobin levels
- Sterile filtered using true pool technology
- Characterized FBS is filtered through sequential, 100 nm pore-size rated filters
- Gamma irradiation further reduces the risk of adventitious viruses present in serum
Specifications
HyClone™ | |
Australia | |
≤ 25 EU/mL | |
Frozen | |
Gamma Irradiated | |
USDA Safety Tested |
100 mL | |
Sterile | |
No | |
≤ 25 mg/dL | |
Filtered through three sequential 100 nm pore size-rated filters | |
Fetal Bovine Serum |