missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Hyclone™ HyClone™ CDM4MAb Media
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Form | Liquid |
| Product Type | Hyclone CDM4MAb Media |
| Without Additives | No Glutamine |
| For Use With (Application) | Production of monoclonal antAntibodies (MAbs) from hybridoma and recombinant myeloma cell lines for research, therapeutic and diagnostic purposes is rapidly growing. |
| Quantity | 500 mL |
| Certifications/Compliance | cGMP |
| Content And Storage | 2 to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction