missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Serum Albumin (BSA)

Product Code. 12330023
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 12330023

Brand: Cytiva SH30574.02

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Support cell culture growth with HyClone™ Bovine Serum Albumin (BSA).

  • pH 7.0
  • Lyophilized powder
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Packaged in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage 2° to 8° C
Form Lyophilized Powder
Product Type BSA
Sterility Non-sterile
Endotoxin Level ≤ 10 EU/mg
pH 6.5 to 7.5
Purity or Quality Grade Cell Culture Grade
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.