missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Calf Serum, US Origin
A cost-effective alternative to Fetal Bovine Serum (FBS)
£8.00 - £54.25
Specifications
Content And Storage | ≤-10°C |
---|---|
Age | 16 to 22 weeks |
Country of Origin | United States |
Sterilization Method | Sequential-filtered, 100 nm |
Source | Formula-Fed Veal Calves |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16655735
|
Cytiva
SH30072.01NI |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16665735
|
Cytiva (Formerly GE Healthcare Life Sciences)
SH30072.02NI |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16675735
|
Cytiva
SH30072.03NI |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Collected by venipuncture from formula-fed veal calves
- Age of animal at time of collection approximately 16-24 weeks
- Complete traceability back to original source
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
- Sourced in the USA
Specifications
≤-10°C | |
United States | |
Formula-Fed Veal Calves |
16 to 22 weeks | |
Sequential-filtered, 100 nm | |
Virus Panel Testing According to 9 CFR 113.53 |