missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline, ES Cell Qualified
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
Description
Support growth of embryonic cell culture growth with HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline.
Specifications
Specifications
| Certifications/Compliance | cGMP and ISO 91:2 Guidelines |
| Content And Storage | 15 to 30°C |
| Concentration | 1 X |
| Product Type | DPBS |
| Sterilization Method | 0.1 μm Sterile-filtered |
| Without Additives | No Calcium, No Magnesium |
| Endotoxin Level | =1.0EU/mL |
| pH | 7.0 to 7.2 |
| Product Line | AdvanceSTEM ES Qualified |
| Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction