missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ ActiCHO™ SM Powdered Media

Product Code. 17275896
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
This item is not returnable. View return policy

Product Code. 17275896

Brand: Cytiva SH31029.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrolysate-free and animal-derived component-free (ADCF)

ActiCHO SM powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.

  • Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides or growth factors in ActiCHO SM medium.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Sterility Non-sterile
With Additives Poloxamer 188
Without Additives No Glutamine, No Insulin
Quantity 1 L
Content And Storage 2 to 8°C, away from light and moisture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.