missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ ActiCHO™ P Powdered Media

Product Code. 17245896
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17245896 5 L 5 L
17255896 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17245896 Supplier Cytiva Supplier No. SH31025.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrolysate-free and animal-derived component-free (ADCF)

ActiCHO P powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.

  • Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides, or growth factors in ActiCHO P medium.
TRUSTED_SUSTAINABILITY

Especificaciones

Form Powder
Sterility Non-sterile
With Additives Poloxamer 188
Without Additives No Glutamine, No Insulin
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.