missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ HybridRack™ Pipet Tip, Extended Length, Maxymum Recovery Surface
GREENER_CHOICE

Product Code. 30090431 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
300 μL
1000 μL
Color:
Blue
Green
Red
Unit Size:
Pack of 960
Product Code. Volume (Metric) Color unitSize
30090431 10 μL Red Pack of 960
30090438 1000 μL Blue Pack of 960
30090441 300 μL Green Pack of 960
3 options
This item is not returnable. View return policy

Product Code. 30090431

Brand: Axygen™ ERF10XTLRS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.

The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.

  • The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
  • The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
  • The paperboard base can be easily recycled through most local curbside recycling programs.
  • All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
  • The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
  • The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.
TRUSTED_SUSTAINABILITY

Specifications

Description Maxymum Recovery Surface
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Red
Autoclavable Not Autoclavable
Format Rack
Tip Style Extended Length
Product Type Pipet Tip
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.