missing translation for 'onlineSavingsMsg'
Learn More

Hyamine Solution 5% (w/v), SLR, Extra Pure, Fisher Chemical

Product Code. 12877182 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12877182 1 L Amber glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12877182 Supplier Fisher Chemical Supplier No. J/9016/15

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121-54-0
Packaging Amber glass bottle
Molecular Formula C27H42ClNO2
MDL Number 11742
Quantity 1 L
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
Molecular Weight (g/mol) 448.088
PubChem CID 8478
ChEBI CHEBI:31264
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.