missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronidase 1/HYAL1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33459-20ul
This item is not returnable.
View return policy
Description
Hyaluronidase 1/HYAL1 Monoclonal antibody specifically detects Hyaluronidase 1/HYAL1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Hyaluronidase 1/HYAL1 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| EC 3.2.1.35, FUS2, Hyal-1, hyaluronidase 1, hyaluronoglucosaminidase 1, Hyaluronoglucosaminidase-1, luCa-1, LUCA1hyaluronidase-1, Lung carcinoma protein 1, MGC45987, NAT6, plasma hyaluronidase, tumor suppressor LUCA-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-435 of human Hyaluronidase 1/HYAL1.2).,, Sequence:, FRGPLLPNRPFTTVWNANTQWCLERHGVDVDVSVFDVVANPGQTFRGPDMTIFYSSQLGTYPYYTPTGEPVFGGLPQNASLIAHLARTFQDILAAIPAPDFSGLAVIDWEAWRPRWAFNWDTKDIYRQRSRALVQAQHPDWPAPQVEAVAQDQFQGAARAWMAGTLQLGRALRPRGLWGFYGFPDCYNYDFLSPNYTGQCPSGIRAQNDQLGWLWGQSRALYPSIYMPAVLEGTGKSQMYVQHRVAEAFRVAVAAGDPNLPVLPYVQIFYDTTNHFLPLDELEHSLGESAAQGAAGVVLWVSWENTRTKESCQAIKEYMDTTLGPFILNVTSGALLCSQALCSGHGRCVRRTSHPKALLLLNPASFSIQLTPGGGPLSLRGALSLEDQAQMAVEFKCRCYPGWQAPWCERKSMW | |
| 20 μL | |
| Immunology, Inflammation | |
| 3373 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction