missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronic acid sodium salt, Streptococcus equi, 91%

Product Code. 15488579
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488579 500 mg 500mg
15480687 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488579 Supplier Thermo Scientific Alfa Aesar Supplier No. J66993.ME
  • ONLINE EXCLUSIVE PRICE
    £47.63
    £38.65 / 500mg
    Save £8.98
    18% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hyaluronic acid sodium salt, CAS # 9067-32-7, is an acidic mucopolysaccharide produced by the bacterium Streptococcus zooepidemicus that is abundant in the mammalian extracellular matrix. | CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol

Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hyaluronic acid sodium salt is a component of the extracellular matrix that is potentially useful for soft tissue augmentation procedures

Application

• Hyaluronic acid sodium salt may be used in formulations of non-animal gels used in soft tissue augmentation studies
• It can be used in laboratory development of artificial synovial fluid
• It can also be used to study its role in oxidative stress and tumor progression

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9067-32-7
Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.30
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

Type Hyaluronic Acid Sodium Salt
Physical Form Powder
Color White
Quantity 500 mg
Merck Index 14,4757
Solubility Information Soluble in water
Percent Purity 91%
Chemical Name or Material Hyaluronic acid sodium salt, Streptococcus equi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.