missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronic acid sodium, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Hyaluronic acid sodium (Sodium hyaluronate) is a biopolymer composed of repeating units of disaccharides with various applications.
Specifications
Specifications
| Chemical Name or Material | Hyaluronic acid sodium |
| Color | White |
| CAS | 9067-32-7 |
| Physical Form | Solid |
| Purity Grade Notes | Research |
| Synonym | Sodium hyaluronate |
| Solubility Information | H2O : 3.33 mg/mL (Need ultrasonic) ∣DMSO : 1 mg/mL (ultrasonic and warming and heat to 80°C) |
| Quantity | 50 mg |
| SMILES | CCC1[C@@H](O)[C@@H](O)O[C@@H](O[C@H]2[C@H](C(O[Na])=O)OC(OC)[C@@H](O)[C@@H]2O)[C@@H]1NC(C)=O.[n] |
| For Use With (Application) | Cancer-programmed cell death |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction