missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronic acid, MedChemExpress

Product Code. 16441436
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16441436

Brand: Medchem Express HYB0633A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hyaluronic acid (corn fermented) is a biopolymer composed of repeating units of disaccharides with various applications.

Hyaluronic acid (corn fermented) is a biopolymer composed of repeating units of disaccharides with various applications.

Specifications

Chemical Name or Material Hyaluronic acid
Color White
CAS 9004-61-9
Physical Form Solid
Purity Grade Notes Research
Molecular Formula (C14H21NO11)n
Synonym Hyaluronate
Solubility Information H2O : 14.29 mg/mL (33.12 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.32 mM; ultrasonic and warming and heat to 80°C)
Quantity 200 mg
SMILES CCC1[C@@H](O)[C@@H](O)O[C@@H](O[C@H]2[C@H](C(O)=O)OC(OC)[C@@H](O)[C@@H]2O)[C@@H]1NC(C)=O.[n]
Molecular Weight (g/mol) 431.49
For Use With (Application) Cancer-programmed cell death
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.