missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronan Synthase 3/HAS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35352-20ul
This item is not returnable.
View return policy
Description
Hyaluronan Synthase 3/HAS3 Polyclonal antibody specifically detects Hyaluronan Synthase 3/HAS3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Hyaluronan Synthase 3/HAS3 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| EC 2.4.1.212, HA synthase 3, hyaluronan synthase 3, Hyaluronate synthase 3, Hyaluronic acid synthase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 67-281 of human Hyaluronan Synthase 3/HAS3 (NP_619515.1).,, Sequence:, HRRMRRAGQALKLPSPRRGSVALCIAAYQEDPDYLRKCLRSAQRISFPDLKVVMVVDGNRQEDAYMLDIFHEVLGGTEQAGFFVWRSNFHEAGEGETEASLQEGMDRVRDVVRASTFSCIMQKWGGKREVMYTAFKALGDSVDYIQVCDSDTVLDPACTIEMLRVLEEDPQVGGVGGDVQPPGKGMAVEDDQVQAAQVRATEAWSVHQRHVSREQ | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3038 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction