missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronan synthase 2 Antibody (4E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£179.00 - £461.00
Specifications
| Antigen | Hyaluronan synthase 2 |
|---|---|
| Clone | 40000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605215
|
Novus Biologicals
NBP2-37446-0.025ml |
0.025 mL |
£179.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18103539
|
Novus Biologicals
NBP2-37446 |
0.1 mL |
£461.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hyaluronan synthase 2 Monoclonal specifically detects Hyaluronan synthase 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Hyaluronan synthase 2 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 3037 | |
| Purified recombinant fragment of human HAS2 (AA: 67-170) expressed in E. Coli. | |
| Primary | |
| 63.5 kDa |
| 40000000 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Mouse | |
| Human | |
| EC 2.4.1.212, HA synthase 2, hyaluronan synthase 2, Hyaluronate synthase 2, Hyaluronic acid synthase 2, MGC126241, MGC126242 | |
| HAS2 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title