missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronan synthase 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£171.00 - £393.00

Specifications

Antigen Hyaluronan synthase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18366026
View Documents
Novus Biologicals
NBP3-03814-20UL
20 μL
£171.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18382185
View Documents
Novus Biologicals
NBP3-03814-100UL
100 μL
£393.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hyaluronan synthase 1 Polyclonal specifically detects Hyaluronan synthase 1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Hyaluronan synthase 1
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
3036
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human, Mouse
HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1
Recombinant fusion protein containing a sequence corresponding to amino acids 80-390 of human Hyaluronan synthase 1 (NP_001514.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato