missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HYAL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550427
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total HYAL3 protein.
This gene encodes a member of the hyaluronidase family. Hyaluronidases are endoglycosidase enzymes that degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. The regulated turnover of hyaluronan plays a critical role in many biological processes including cell proliferation, migration and differentiation. The encoded protein may also play an important role in sperm function. This gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression, and the expression of specific transcript variants may be indicative of tumor status. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and some isoforms may lack hyaluronidase activity. This gene overlaps and is on the same strand as N-acetyltransferase 6 (GCN5-related), and some transcripts of each gene share a portion of the first exon.
Specifications
| HYAL3 | |
| Polyclonal | |
| Unconjugated | |
| HYAL3 | |
| Hyal3; hyal-3; Hyaluronidase; hyaluronidase 3; hyaluronidase-3; hyaluronoglucosaminidase 3; hyaluronoglucosaminidase-3; Hyl3; LUCA14; LUCA3; LuCa-3; Lung carcinoma protein 3; Minna14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8372 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O43820 | |
| HYAL3 | |
| Fusion protein corresponding to a region derived from internal residues of human hyaluronoglucosaminidase 3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction