missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYAL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524223
Code : LD
Additional Details : Weight : 0.01000kg
Beschreibung
This gene encodes a weak acid-active hyaluronidase. The encoded protein is similar in structure to other more active hyaluronidases. Hyaluronidases degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan and fragments of hyaluronan are thought to be involved in cell proliferation, migration and differentiation. Although it was previously thought to be a lysosomal hyaluronidase that is active at a pH below 4, the encoded protein is likely a GPI-anchored cell surface protein. This hyaluronidase serves as a receptor for the oncogenic virus Jaagsiekte sheep retrovirus. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. This gene encodes two alternatively spliced transcript variants which differ only in the 5' UTR.Spezifikation
HYAL2 | |
Polyclonal | |
Unconjugated | |
HYAL2 | |
AI256841; AU020858; AW555733; HYAL2; Hyal-2; Hyaluronidase; hyaluronidase 2; hyaluronidase-2; hyaluronoglucosaminidase 2; hyaluronoglucosaminidase-2; LUCA2; LuCa-2; lung carcinoma protein 2; lysosomal hyaluronidase; PH20 homolog; PH-20 homolog | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8692 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
Q12891 | |
HYAL2 | |
KLH conjugated synthetic peptide between 385-412 amino acids from the C-terminal region of human HYAL2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |