missing translation for 'onlineSavingsMsg'
Learn More

HYAL2, Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse Polyclonal Antibody

Brand:  Abnova H00008692-B02P.50ug

Product Code. 16159055

  • £303.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MRAGPGPTVTLALVLAVAWAMELKPTAPPIFTGRPFVVAWDVPTQDCGPRLKVPLDLNAFDVQASPNEGFVNQNITIFYRDRLGLYPRFDSAGRSVHGGVPQNVSLWAHRKMLQKRVEHYIRTQESAGLAVIDWEDWRPVWVRNWQDKDVYRRLSRQLVASRHPDWPPDRIVKQAQYEFEFAAQQFMLETLRYVKAVRPRHLWGFYLFPDCYNHDYVQNWESYTGRCPDVEVARNDQLAWLWAESTALFPSVYLDETLASSRHGRNFVSFRVQEALRVARTHHANHALPVYVFTRPTYSRRLTGLSEMDLISTIGESAALGAAGVILWGDAGYTTSTETCQYLKDYLTRLLVPYVVNVSWATQYCSRAQCHGHGRCVRRNPSASTFLHLSTNSFRLVPGHAPGEPQLRPVGELSWADIDHLQTHFRCQCYLGWSGEQCQWDHRQAAGGASEAWAGSHLTSLLALAALAFTWTL
Specifications

Specifications

HYAL2
Polyclonal
In 1x PBS, pH 7.4
DQ893838.2
HYAL2
HYAL2 (ABM84764.1, 1 a.a. ∼ 473 a.a) full-length human protein.
RUO
8692
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Purified
Western Blot
Unconjugated
hyaluronoglucosaminidase 2
LUCA2/LuCa-2
Mouse
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.