missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYAL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25179-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. Hyaluro is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| HYAL1 | |
| Polyclonal | |
| Unconjugated | |
| HYAL1 | |
| HYAL 1, HYAL1, Hyaluronidase 1, hyaluronoglucosaminidase 1, LuCa 1, LUCA1, Lung carcinoma protein 1, NAT6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15586, 3373 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q12794, Q91ZJ9 | |
| HYAL1 | |
| HYAL1 Fusion Protein Ag17943 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction