missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HYAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551686
This item is not returnable.
View return policy
Description
Immunogen sequence: SRALVQAQHP DWPAPQVEAV AQDQFQGAAR AWMAGTLQLG RALRPRGLWG FYGFPDCYNY DFLSPNYTGQ CPSGIRAQND QLGWLWGQSR ALYPSIYMPA VLEGTGKSQM YVQHRVAEAF RVAVAAGDPN LPVLPYVQIF YDTTNHF Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| HYAL1 | |
| Polyclonal | |
| Unconjugated | |
| HYAL1 | |
| Hya1; HYAL1; Hyal-1; Hyaluronidase; hyaluronidase 1; hyaluronidase-1; hyaluronoglucosaminidase 1; hyaluronoglucosaminidase-1; LUCA1; LuCa-1; Lung carcinoma protein 1; MPS9; NAT6; plasma hyaluronidase; tumor suppressor LUCA-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12794 | |
| HYAL1 | |
| Recombinant protein corresponding to Human HYAL1. Recombinant protein control fragment (Product #RP-101871). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction