missing translation for 'onlineSavingsMsg'
Learn More

HYAL1 Mouse anti-Human, Rat, Clone: 1B8B5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66560-1-IG-150UL

 View more versions of this product

Product Code. 16878545

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. Hyaluro is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

HYAL1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12794, Q76HN1
HYAL1
HYAL1 Fusion Protein Ag18183
150 μL
Primary
Human, Rat
Antibody
IgG1
Western Blot
1B8B5
Unconjugated
HYAL1
HYAL 1, HYAL1, Hyaluronidase 1, hyaluronoglucosaminidase 1, LuCa 1, LUCA1, Lung carcinoma protein 1, NAT6
Mouse
Protein G
RUO
3373, 367166
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.