Learn More
Hamilton™ HVXM Large Body Modular Valve Positioner PTFE Valves
Click to view available options
Model:
3-5 model, 3 valves
4-5 model, 4 valves
Unit Size:
Each
Description
- Physically the largest valves
- Even moderately viscous fluids flow freely
- Exceptionally easy to service and modify
- Maximizes flexibility and value
Specifications
Specifications
| Temperature (Metric) Max | 38°C |
| Model | 3-5 model, 3 valves |
| Volume (Metric) | 108.8 μL dead volume |
| Thread Size | 1/4 in.-28 |
| Description | Large body PTFE valve |
| Material | Aluminum (Housing) |