Learn More
Hamilton™ HVXM Large Body Modular Valve Positioner PTFE Valves
Product Code. 11533274
Click to view available options
Model:
3-5 model, 3 valves
4-5 model, 4 valves
Unit Size:
Each
Product Code. 11533274
Brand: Hamilton™ 36758
Only null left
Add to basket
Add to basket
With a larger port internal diameter to allow increased fluid flows.
  • Physically the largest valves
  • Even moderately viscous fluids flow freely
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
Specifications
Temperature (Metric) Max 38°C
Model 3-5 model, 3 valves
Volume (Metric) 108.8 μL dead volume
Thread Size 1/4 in.-28
Description Large body PTFE valve
Material Aluminum (Housing)