missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hVEGF-IN-1, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
hVEGF-IN-1, a quinazoline derivative, could specifically bind to the G-rich sequence in the internal ribosome entry site A (IRES-A) and destabilize the G-quadruplex structure. hVEGF-IN-1 binds to the IRES-A (WT) with a Kd of 0.928 μM in SPR experiments. hVEGF-IN-1 could hinder tumor cells migration and repress tumor growth by decreasing VEGF-A protein expression.
Specifications
Specifications
| Chemical Name or Material | hVEGF-IN-1 |
| CAS | 1637443-98-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H43N7O2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (42.97 mM; Need ultrasonic) |
| SMILES | CCN(CC)CCOC(C=C1)=CC=C1NC2=NC(C3=C(NC(CCN4CCN(C)CC4)=O)C=CC=C3)=NC5=CC=CC=C52 |
| Molecular Weight (g/mol) | 581.75 |
| Formula Weight | 581.75 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction