Learn More
Hamilton™ HVC Standard Ceramic Valves
Designed to provide superior performance with salt solutions.
Brand: Hamilton™ 6764-01
Code : NEW
Additional Details : Weight : 0.08800kg
Description
- Rigid ceramic fluid path is resistant to scratching from salt crystals
- Provides superior life and performance
- Exceptionally easy to service and modify
- Maximizes flexibility and value
- Ideal for low pressure applications
Specifications
Stainless Steel (Housing) | |
Ceramic HV valve | |
3-5 Model, 3 Ports, Stainless Steel Housing | |
PSD/2 and 3 Syringe Drive Modules |