missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HUWE1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12808562
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808562 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808562 Supplier Invitrogen™ Supplier No. PA520374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Daudi cell lysate. PA5-20374 can be used with blocking peptide PEP-0491.

SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen HUWE1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HUWE1
Gene Accession No. P51593, Q7TMY8, Q7Z6Z7
Gene Alias 5430439H10Rik; ARF-binding protein 1; ARF-BP1; AU041296; BJ-HCC-24 tumor antigen; C430014N20Rik; C80292; E3 ubiquitin-protein ligase HUWE1; E3Histone; Gm1718; HECT domain protein LASU1; HECT, UBA and WWE domain containing 1; HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase; HECT, UBA and WWE domain-containing protein 1; HectH9; HECT-type E3 ubiquitin transferase HUWE1; Homologous to E6AP carboxyl terminus homologous protein 9; HSPC272; HUWE1; Ib772; Kiaa0312; KIAA1578; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; Mule; RP3-339A18.4; upstream regulatory element binding protein 1; upstream regulatory element-binding protein 1; Ureb1; URE-B1; URE-binding protein 1
Gene Symbols HUWE1
Host Species Rabbit
Immunogen A 19 amino acid peptide from near the carboxy terminus of human ARF-BP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10075, 501546, 59026
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.