missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUWE1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-652JF549
This item is not returnable.
View return policy
Description
HUWE1 Polyclonal specifically detects HUWE1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HUWE1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| arf-bp1, EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| Stem Cell Markers | |
| 10075 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HUWE1 | |
| The immunogen this antibody was made to, maps to a region between residues 2250 and 2300 of human Upstream Regulatory Element Binding Protein 1 using the numbering given in entry NP_113584.3 (GeneID 10075). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction