missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HUS1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103493
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HUS1B.
HUS1Bis most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. HUS1B can interact with the check point protein RAD1 but not with RAD9. Overexpression of HUS1B has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1. The protein encoded by this gene is most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. This protein can interact with the check point protein RAD1 but not with RAD9. Overexpression of this protein has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1.
Specifications
| HUS1B | |
| Polyclonal | |
| Unconjugated | |
| Hus1b | |
| Checkpoint protein HUS1B; hHUS1B; HUS1 checkpoint clamp component B; HUS1 checkpoint homolog b; HUS1 checkpoint homolog b (S. pombe); Hus1 homolog b (S. pombe); Hus1b; MGC126746; MGC126748; mHUS1B; RP11-532F6.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 135458, 691382 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NHY5 | |
| Hus1b | |
| A synthesized peptide derived from human HUS1B(Accession Q8NHY5), corresponding to amino acid residues F66-V116. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction