missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HUS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597849
This item is not returnable.
View return policy
Description
HUS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by this gene is a component of an evolutionarily conserved, genotoxin-activated checkpoint complex that is involved in the cell cycle arrest in response to DNA damage. This protein forms a heterotrimeric complex with checkpoint proteins RAD9 and RAD1.
Specifications
| HUS1 | |
| Polyclonal | |
| Unconjugated | |
| HUS1 | |
| Checkpoint protein HUS1; hHUS1; Hus1; HUS1 checkpoint clamp component; HUS1 checkpoint homolog; HUS1 checkpoint homolog (S. pombe); Hus1 homolog; Hus1 homolog (S. pombe); HUS1 protein; hus1+-like protein; mHUS1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 3364 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O60921 | |
| HUS1 | |
| Recombinant Human Checkpoint protein HUS1 protein (2-280AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction