missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HUS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555417
This item is not returnable.
View return policy
Description
Immunogen sequence: NHFTRISNMI AKLAKTCTLR ISPDKLNFIL CDKLANGGVS MWCELEQENF FNEFQMEGVS AENNEIYLEL TSENLSRASK TAQNARALKI KLTNKHFPCL TVSVELLSMS SSSRIVTHD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
The protein encoded by this gene is a component of an evolutionarily conserved, genotoxin-activated checkpoint complex that is involved in the cell cycle arrest in response to DNA damage. This protein forms a heterotrimeric complex with checkpoint proteins RAD9 and RAD1.
Specifications
| HUS1 | |
| Polyclonal | |
| Unconjugated | |
| HUS1 | |
| Checkpoint protein HUS1; hHUS1; Hus1; HUS1 checkpoint clamp component; HUS1 checkpoint homolog; HUS1 checkpoint homolog (S. pombe); Hus1 homolog; Hus1 homolog (S. pombe); HUS1 protein; hus1+-like protein; mHUS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3364 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60921 | |
| HUS1 | |
| Recombinant protein corresponding to Human HUS1. Recombinant protein control fragment (Product #RP-89479). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction