missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HUS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522306
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, C8D30. Predicted reactivity: Mouse (85%), Rat (86%), Pig (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a component of an evolutionarily conserved, genotoxin-activated checkpoint complex that is involved in the cell cycle arrest in response to DNA damage. This protein forms a heterotrimeric complex with checkpoint proteins RAD9 and RAD1.
Specifications
| HUS1 | |
| Polyclonal | |
| Unconjugated | |
| HUS1 | |
| Checkpoint protein HUS1; hHUS1; Hus1; HUS1 checkpoint clamp component; HUS1 checkpoint homolog; HUS1 checkpoint homolog (S. pombe); Hus1 homolog; Hus1 homolog (S. pombe); HUS1 protein; hus1+-like protein; mHUS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15574, 3364, 498411 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.71 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O60921, Q8BQY8 | |
| HUS1 | |
| Recombinant protein encompassing a sequence within the center region of human HUS1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction