Learn More
Invitrogen™ HuR Recombinant Rabbit Monoclonal Antibody (23GB5815), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553928
 View more versions of this product
Product Code. 30088773
  • £485.00 / 100µL
Description
The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HuR
Recombinant Monoclonal
Unconjugated
ELAVL1
2410055N02Rik; ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; ELAV like RNA binding protein 1; ELAV like RNA binding protein 1a; ELAV1; Elavl1; elavl1 protein; elavl1a; elav-like generic protein; ELAV-like protein 1; ELAV-like protein 1; ELAV-like protein 1a; elav-related A; Elra; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-related A; etID309757.2; fa25f08; fb59a02; fc60g02; Hu antigen R; Hua; HU-antigen A; hu-antigen R; Human antigen R; HuR; melG; ribonucleoprotein; W91709; wu:fa25f08; wu:fb59a02; wu:fc60g02; zHuA
Rabbit
Affinity chromatography
RUO
15568, 1994, 363854
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5815
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
B5DF91, P70372, Q15717
ELAVL1
A synthesized peptide derived from human HuR/ELAVL1 (52-96AA).
100 μL
Primary
Human, Mouse, Rat