missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HuR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583813
This item is not returnable.
View return policy
Description
Immunogen sequence: MSNGYEDHMA EDCRGDIGRT NLIVN Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.
Specifications
| HuR | |
| Polyclonal | |
| Unconjugated | |
| ELAVL1 | |
| 2410055N02Rik; ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; ELAV like RNA binding protein 1; ELAV like RNA binding protein 1a; ELAV1; Elavl1; elavl1 protein; elavl1a; elav-like generic protein; ELAV-like protein 1; ELAV-like protein 1; ELAV-like protein 1a; elav-related A; Elra; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-related A; etID309757.2; fa25f08; fb59a02; fc60g02; Hu antigen R; Hua; HU-antigen A; hu-antigen R; Human antigen R; HuR; melG; ribonucleoprotein; W91709; wu:fa25f08; wu:fb59a02; wu:fc60g02; zHuA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1994 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15717 | |
| ELAVL1 | |
| Recombinant protein corresponding to Human ELAVL1. Recombinant protein control fragment (Product #RP-102986). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction