missing translation for 'onlineSavingsMsg'
Learn More

Huntingtin Interacting Protein K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-05862-100ul

 View more versions of this product

Product Code. 18623868

  • £255.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Huntingtin Interacting Protein K Polyclonal antibody specifically detects Huntingtin Interacting Protein K in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Huntingtin Interacting Protein K
Polyclonal
Immunohistochemistry-Paraffin 1:200-1:1000
C15orf63, chromosome 15 open reading frame 63, EC 2.1.1.43, EC 2.7.7.6, FLJ20431, HSPC136, Huntingtin yeast partner K, huntingtin-interacting protein K, HYPKHuntingtin interacting protein K
Produced in rabbits immunized with E. coli-derived Human Huntingtin Interacting Protein K fragment.
100 μg
Primary
Human
Purified
Immunohistochemistry (Paraffin)
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
25764
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.