missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huntingtin Interacting Protein K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49177
This item is not returnable.
View return policy
Description
Huntingtin Interacting Protein K Polyclonal antibody specifically detects Huntingtin Interacting Protein K in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Huntingtin Interacting Protein K | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| C15orf63, chromosome 15 open reading frame 63, EC 2.1.1.43, EC 2.7.7.6, FLJ20431, HSPC136, Huntingtin yeast partner K, huntingtin-interacting protein K, HYPKHuntingtin interacting protein K | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TAMSVIGDRRSREQKAKQEREKELAKVTIKKEDLELIMTEMEISRAAAERSLREHMGNVVEALIALT | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 25764 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction