missing translation for 'onlineSavingsMsg'
Learn More

HUNK Antibody (1C4), Novus Biologicals™

Product Code. 18366489 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366489 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366489 Supplier Novus Biologicals Supplier No. H00030811M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HUNK Monoclonal antibody specifically detects HUNK in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HUNK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C4
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055401
Gene Alias B19, EC 2.7.11, EC 2.7.11.1, hormonally up-regulated neu tumor-associated kinase, hormonally upregulated neu tumor-associated kinase, hormonally up-regulated Neu-associated kinase, hormonally upregulated Neu-associated kinase, MAKV, serine/threonine protein kinase MAK-V, Serine/threonine-protein kinase MAK-V
Host Species Mouse
Immunogen HUNK (NP_055401, 615 a.a. ~ 714 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LHPTLVSFAHEDKNSPPKEEGLCCPPPVPSNGPMQPLGSPNCVKSRGRFPMMGIGQMLRKRHQSLQPSADRPLEASLPPLQPLAPVNLAFDMADGVKTQC
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 30811
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.