missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human Transferrin protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
Transferrin is an 80kDA, monomeric glycoprotein that is responsible for binding ferric iron and transporting it throughout the body. It is primarily produced by hepatocytes and is a critical component of iron homeostasis as it delivers free iron to tissue sites where it is absorbed and utilized by the body. Once it reaches a site of absorption, iron-loaded transferrin binds to its corresponding transferrin receptor and taken up by the cell through endocytosis, after which the iron is disassociated, and free transferrin is released back into the bloodstream at a turnover rate of around 10 times per day. In addition to iron homeostasis, transferrin also plays an important role in the innate immune system. Downregulation of transferrin receptor expression has been linked to the reduced ability of intracellular pathogens to access a cell’s iron supply. Measurement of transferrin levels in the blood can also serve as a clinical marker for possible iron deficiency (PMID: 30422523, 22033148).
Specifications
Specifications
| Accession Number | P02787 |
| Formulation | 1 x PBS |
| Gene ID (Entrez) | 7018 |
| Molecular Weight (g/mol) | 70 kDa |
| Quantity | 1000 μg |
| Source | HEK293 |
| Storage Requirements | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Endotoxin Concentration | <0.1 EU/μg |
| Specific Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction