missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ HumanKine™ Recombinant Human Thrombin (Coagulation Factor II) Protein

Product Code. 16801436 Shop All Proteintech Products
Click to view available options
Quantity:
10 x 1000 μg
1000 μg
5000 μg
Unit Size:
1000µg
5000µg
Pack of 10
This item is not returnable. View return policy

Product Code. 16801436

Brand: Proteintech™ HZ30105000UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells

  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity

Specifications

Gene ID (Entrez) 2147
Molecular Weight (g/mol) 36 kDa, glycosylated
Quantity 5000 μg
Species Human
Content And Storage This protein is stable for 1 year from the date of receipt if stored between -20 to -80°C. Repeated freeze thaw cycles should be avoided.
Expression System Human embryonic kidney 293 (HEK293) human cell expression system
Form Lyophilized
Purity or Quality Grade > 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.