missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ HumanKine™ Recombinant Human R-Spondin-I Protein

Product Code. 17711499 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17711499 10 μg 10µg
17701499 100 μg 100µg
17791489 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17711499 Supplier Proteintech™ Supplier No. HZ132810UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R-Spondin 1 plays several roles in embryonic development including sex phenotype reversal, female sex determination, and ovarian differentiation.

R-Spondin 1 is an approximately 40kDA, glycosylated protein that serves as a major agonist of the Wnt signaling pathway. It regulates the turnover of the LRP6 co-receptor by preventing its internalization by DKK-1. It also promotes stem cell turnover in the mammary glands, intestine, colon, and kidneys. Increased R-Spondin 1 expression has been linked to growth and migration of ovarian cancer cells as well as decreased sensitivity of gliomas to radiation-based therapies (PMID: 17804805, 22439850, 32749219, 30572097).

Specifications

Gene ID (Entrez) 284654
Quantity 10 μg
Source Human
Endotoxin Concentration <1 EU/μg
Specific Reactivity Human
Mass 40 kDa
Form Lyophilized
Purity or Quality Grade >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.