missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IL-8 protein

Product Code. 17680077 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17680077

Brand: Proteintech HZ131810UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HumanKine™ recombinant protein

IL-8 is a chemoattractant cytokine that signals through the CXCR1 and CXCR2 receptors to attract and activate neutrophils to sites of inflammation. It is secreted by multiple types of cells, including monocytes, neutrophils, tumor, epithelial, endothelial, and fibroblasts. IL-8 signaling activates downstream pathways that promote cell survival, proliferation, and cell invasiveness. As such, IL-8 has been linked to the promotion of angiogenesis, immunosuppressive cell recruitment, and EMT transition in several cancer types. Recent studies also indicate that IL-8 can serve as a marker for COVID-19 disease progression (PMID: 22235381, 18980965, 26548401, 33584733).

Specifications

Accession Number P10145
Formulation 1 x PBS
Gene ID (Entrez) 3576
Molecular Weight (g/mol) 12 kDa
Quantity 10 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <0.1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.