missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human IL-5 protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
IL-5 is a 4-α-helix, pleiotropic cytokine that regulates the activities of B cells, eosinophils, and basophils. It is produced mostly by CD4+ T cells, eosinophils, and basophils, and signals through its specific receptor (IL-5R) on target cells to activate downstream JAK/STAT, Btk, and Ras/ERk pathways. Due to its pleiotropic effects, IL-5 plays a role in both innate and adaptive immune responses. It is a strong promoter of eosinophil differentiation, migration, maturation, and survival. Additionally, IL-5 is also capable of stimulating activated B cells to differentiate into antibody-secreting cells. Elevated IL-5 levels are strongly correlated with eosinophilia, which is a hallmark feature of allergy, asthma, and hypersensitivity (PMID: 21986312, 22174445, 19819937, 16895557)
Specifications
Specifications
| Accession Number | P05113 |
| Formulation | 1 x PBS |
| Gene ID (Entrez) | 3567 |
| Molecular Weight (g/mol) | 35-38 kDa |
| Quantity | 10 μg |
| Source | HEK293 |
| Storage Requirements | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Endotoxin Concentration | <0.1 EU/μg |
| Specific Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction