missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human IL-34 protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
IL-34 is a 6-α-helix, proinflammatory cytokine that serves as a ligand for colony-stimulating factor-1 receptor (CSF1R). It is expressed most abundantly by spleen cells. It primarily stimulates human peripheral blood monocyte proliferation and macrophage colony formation. It is also required for Langerhans cell and microglia development. IL-34 has been shown to play a protective role in cancer and Alzheimer’s disease, but it can promote the progression of several autoimmune, inflammatory, and metabolic diseases. As such IL-34 could be used as a biomarker to mark the progression of these diseases (PMID: 22579672, 27577879, 31940023).
Specifications
Specifications
| Accession Number | Q6ZMJ4 |
| Formulation | 50 mM Acetate pH 4.5 + 75 mM NaCl, See Certificate of Analysis for details |
| Gene ID (Entrez) | 146433 |
| Molecular Weight (g/mol) | 68 and 38 kDa |
| Quantity | 1000 μg |
| Source | HEK293 |
| Storage Requirements | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Endotoxin Concentration | <1 EU/μg |
| Specific Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction