missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human IL-22 protein
Tienda Proteintech Productos
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Tamaño de la unidad:
1000µg
100µg
10µg
Specifications
Specifications
| Accession Number | Q9GZX6 |
| Formulation | 1 x PBS, See Certificate of Analysis for details |
| Gene ID (Entrez) | 50616 |
| Molecular Weight (g/mol) | 25-40 kDa, dimer, glycosylated |
| Quantity | 10 μg |
| Source | HEK293 |
| Storage Requirements | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Endotoxin Concentration | <1.0 EU/μg |
| Specific Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction