missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human DKK-1 protein

Product Code. 17670067 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17670067

Brand: Proteintech HZ13141000UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HumanKine™ recombinant protein

DKK-1 (Dickkopf WNT signaling pathway inhibitor 1) is a member of the DKK protein family. Other members of this family include DKK-2, DKK-3, and DKK-4. The DKK family proteins share 35-40 % homology between various members. Recombinant human DKK-1 expressed in HEK293 cells is a 35-45kDa glycosylated protein. It is a secreted protein that functions as a negative regulator of Wnt/β-catenin signaling pathway. Wnt/ β-catenin signaling pathway involves binding of the Wnt ligands to the seven-transmembrane receptor, Frizzled, and the coreceptor lipoprotein-related protein 5 and 6 (LRP5/6). DKK1 forms a ternary complex with LRP5/6 and another receptor, Kremen, followed by endocytosis of this complex and removal of LRP5/6 from the cell surface. It has been suggested that DKK-1 permits anterior development by inhibiting Wnt/ β-catenin signaling, which is essential for posterior patterning in vertebrates. The balance between Wnt signaling and DKK-1 inhibition is critical for bone formation and homeostasis and insufficient or excess DKK-1 activity in bone results in increased or decreased bone density, respectively. (PMID: 17143291; 12050670; 10570958)

Specifikationer

Accession Number O94907
Formulation 1 x PBS, See Certificate of Analysis for details
Gene ID (Entrez) 22943
Molecular Weight (g/mol) 35-38 kDa
Quantity 1000 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Activity 7-70 ng/mL EC50
Form Lyophilized
Purity or Quality Grade >95%
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.