missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Humanin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141325
This item is not returnable.
View return policy
Description
Suggested positive control: recombinant protein.
Humanin is a novel 24 A. A polypeptide identified in Alzheimer's disease-affected brain. It is 99% homologous to mitochondrial 16s ribosomal RNA and functions as a novel neuroprotective and cytoprotective factor. It specifically inhibits neuronal cell death induced by beta-amyloids and Alzheimer's disease causing FAD genes by competitively inhibiting access of FPRL1 receptor to beta-amyloids. It acts as an anti-apoptotic factor by binding to Bcl-2/Bax family of pro-apoptotic factors including Bax, Bak, Bid and BimEL and inhibits their pro-apoptotic activity. It activates JAK/STAT signaling pathway and is regulated by TRIM11. It interacts with IGFBP-3 and specifically blocks IGFBP-3 induced cell death in glioblastoma cells. Humanin is also expressed in colon and testis.
Specifications
| Humanin | |
| Polyclonal | |
| Unconjugated | |
| MT-RNR2 | |
| BCL2L4; HN1; Humanin-like 1; humanin-like protein 1; mitochondrially encoded 16S RNA; MTRNR2; MT-RNR2; MTRNR2L1; MT-RNR2-like 1; MTRNR2-like 1; MT-RNR2-like protein 1; RNR2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Conc. Not Determined | |
| Whole serum with 50% glycerol and 0.025% sodium azide | |
| Q8IVG9 | |
| MT-RNR2 | |
| The full length peptide sequence from the human MT-RNR2 protein. | |
| RUO | |
| 4550 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction